Assay Details
| Gene Symbol: Tnni1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_017184.1 | NP_058880.1 | 3 - 4 | 100 | 60 | |
| XM_006249862.2 | XP_006249924.1 | 2 - 3 | 1607 | 60 | ||
| XM_006249864.1 | XP_006249926.1 | 2 - 3 | 186 | 60 | ||
| XM_006249865.2 | XP_006249927.1 | 5 - 6 | 229 | 60 | ||
| XM_006249866.2 | XP_006249928.1 | 5 - 6 | 160 | 60 | ||
| XM_006249868.2 | XP_006249930.1 | 3 - 4 | 195 | 60 | ||
| XM_006249869.2 | XP_006249931.1 | 3 - 4 | 119 | 60 | ||
| GenBank mRNA | FQ214889.1 | - | 5 - 6 | 207 | 60 | |
| FQ215520.1 | - | 5 - 6 | 182 | 60 | ||
| FQ215574.1 | - | 5 - 6 | 180 | 60 | ||
| FQ215749.1 | - | 5 - 6 | 180 | 60 | ||
| FQ216094.1 | - | 5 - 6 | 180 | 60 | ||
| FQ216822.1 | - | 5 - 6 | 180 | 60 | ||
| FQ217411.1 | - | 5 - 6 | 180 | 60 | ||
| J04993.1 | - | 3 - 4 | 100 | 60 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_006249868.2, XM_006249869.2 transcripts.
Set Membership: |
|

Tnni1
Genomic Map