Assay Details
| Gene Symbol: Adh4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_017270.2 | NP_058966.2 | 7 - 8 | 1011 | 69 | |
| GenBank mRNA | BC127504.1 | - | 7 - 8 | 1011 | 7325543 | 69 |
| X90710.1 | - | 7 - 8 | 1037 | 69 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Adh4
Genomic Map