Assay Details
| Gene Symbol: Psma4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_017281.1 | NP_058977.1 | 7 - 8 | 604 | 95 | |
| GenBank mRNA | FQ212536.1 | - | 3 - 4 | 237 | 95 | |
| FQ213330.1 | - | 3 - 4 | 237 | 95 | ||
| FQ215861.1 | - | 3 - 4 | 237 | 95 | ||
| FQ216011.1 | - | 3 - 4 | 237 | 95 | ||
| FQ216241.1 | - | 3 - 4 | 237 | 95 | ||
| FQ220234.1 | - | 3 - 4 | 236 | 95 | ||
| FQ220280.1 | - | 3 - 4 | 237 | 95 | ||
| FQ226777.1 | - | 3 - 4 | 237 | 95 | ||
| FQ234380.1 | - | 3 - 4 | 237 | 95 | ||
| X53304.1 | - | 7 - 8 | 604 | 95 | ||
| X55986.1 | - | 7 - 8 | 560 | 95 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Psma4
Genomic Map