Assay Details
| Gene Symbol: Sepw1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_013027.3 | NP_037159.4 | 2 - 3 | 141 | 99 | |
| GenBank mRNA | BC087625.1 | - | 2 - 3 | 117 | 7316562 | 99 |
| BI279050.1 | - | 99 | ||||
| DN931873.1 | - | 2 - 3 | 163 | 99 | ||
| FQ212381.1 | - | 2 - 3 | 127 | 99 | ||
| FQ212407.1 | - | 2 - 3 | 121 | 99 | ||
| FQ215130.1 | - | 2 - 3 | 120 | 99 | ||
| FQ215391.1 | - | 2 - 3 | 122 | 99 | ||
| FQ216964.1 | - | 2 - 3 | 117 | 99 | ||
| FQ217212.1 | - | 2 - 3 | 122 | 99 | ||
| FQ217426.1 | - | 2 - 3 | 123 | 99 | ||
| FQ217557.1 | - | 2 - 3 | 123 | 99 | ||
| FQ217567.1 | - | 2 - 3 | 123 | 99 | ||
| FQ217929.1 | - | 2 - 3 | 123 | 99 | ||
| FQ221681.1 | - | 2 - 3 | 123 | 99 | ||
| FQ223747.1 | - | 2 - 3 | 123 | 99 | ||
| FQ224556.1 | - | 2 - 3 | 127 | 99 | ||
| FQ224570.1 | - | 2 - 3 | 123 | 99 | ||
| FQ224606.1 | - | 2 - 3 | 120 | 99 | ||
| U25264.1 | - | 2 - 3 | 108 | 99 | ||
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Sepw1
Genomic Map