Assay Details
| Gene Symbol: Ptger4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_006231996.2 | XP_006232058.1 | 1 - 2 | 2507 | 74 | |
| GenBank mRNA | D28860.1 | - | 1 - 2 | 118 | 74 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the D28860.1, XM_006231996.2 transcripts.
Set Membership: |
|

Ptger4
Genomic Map