Assay Details
| Gene Symbol: Cdc42 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_171994.4 | NP_741991.3 | 3 - 4 | 248 | 90 | |
| XM_008764286.1 | XP_008762508.1 | 3 - 4 | 753 | 90 | ||
| XM_008764287.1 | XP_008762509.1 | 3 - 4 | 328 | 90 | ||
| GenBank mRNA | AF491841.1 | - | 2 - 3 | 241 | 90 | |
| BC060535.1 | - | 3 - 4 | 248 | 6920523 | 90 | |
| FQ213007.1 | - | 3 - 4 | 90 | |||
| FQ225045.1 | - | 3 - 4 | 247 | 90 | ||
| FQ225541.1 | - | 3 - 4 | 360 | 90 | ||
| FQ225973.1 | - | 3 - 4 | 360 | 90 | ||
| FQ226943.1 | - | 3 - 4 | 243 | 90 | ||
| FQ227167.1 | - | 3 - 4 | 247 | 90 | ||
| FQ227499.1 | - | 3 - 4 | 247 | 90 | ||
| FQ227894.1 | - | 3 - 4 | 247 | 90 | ||
| FQ228149.1 | - | 3 - 4 | 360 | 90 | ||
| FQ230725.1 | - | 3 - 4 | 256 | 90 | ||
| FQ231089.1 | - | 3 - 4 | 247 | 90 | ||
| FQ231482.1 | - | 3 - 4 | 256 | 90 | ||
| FQ231846.1 | - | 3 - 4 | 360 | 90 | ||
| FQ232179.1 | - | 3 - 4 | 247 | 90 | ||
| FQ232295.1 | - | 3 - 4 | 256 | 90 | ||
| FQ232459.1 | - | 3 - 4 | 247 | 90 | ||
| FQ233586.1 | - | 3 - 4 | 258 | 90 | ||
| FQ234217.1 | - | 3 - 4 | 247 | 90 | ||
| FQ234639.1 | - | 3 - 4 | 256 | 90 | ||
| FQ234830.1 | - | 3 - 4 | 256 | 90 | ||
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Cdc42
Genomic Map