Assay Details
| Gene Symbol: Six3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_023990.1 | NP_076480.1 | 1 - 2 | 1188 | 156 | |
| GenBank mRNA | AB030833.1 | - | 1 - 2 | 1188 | 156 | |
More Information
Updated Mapping Information
Set Membership: |
|

Six3
Genomic Map