Assay Details
| Gene Symbol: Cd2ap | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_181475.2 | NP_852140.1 | 11 - 12 | 1279 | 123 | |
| GenBank mRNA | AY205155.1 | - | 11 - 12 | 1279 | 123 | |
More Information
Updated Mapping Information
Set Membership: |
|
Matching Ambion siRNAs:
Gene Ontology Categories:
Function(s)
Process(es)
mitotic nuclear division
vesicle organization
single organismal cell-cell adhesion
cell migration
negative regulation of transforming growth factor beta1 production
proteasome-mediated ubiquitin-dependent protein catabolic process
regulation of receptor-mediated endocytosis
cell division
positive regulation of protein localization to nucleus
regulation of actin cytoskeleton reorganization
vascular endothelial growth factor receptor binding
protein binding
beta-catenin binding
protein C-terminus binding
SH3 domain binding
protein complex binding
cadherin binding
mitotic nuclear division
vesicle organization
single organismal cell-cell adhesion
cell migration
negative regulation of transforming growth factor beta1 production
proteasome-mediated ubiquitin-dependent protein catabolic process
regulation of receptor-mediated endocytosis
cell division
positive regulation of protein localization to nucleus
regulation of actin cytoskeleton reorganization
vascular endothelial growth factor receptor binding
protein binding
beta-catenin binding
protein C-terminus binding
SH3 domain binding
protein complex binding
cadherin binding
MicroArray Probes:
Agilent
Affymetrix
ABI
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Whole Rat Genome Microarray 4x44K v2 | A_44_P355861 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 355673 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 832035 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 77631 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 229252 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 459685 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 542462 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 214 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 484030 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 5138 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 316173 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 568962 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 596611 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 962053 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 345351 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 323165 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 85511 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 211568 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 739353 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 831139 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 870612 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 536715 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 342222 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 323102 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 315409 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 636137 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 33665 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 281680 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 686896 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 134345 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 503545 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 198554 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 421287 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 55774 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 589504 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 471581 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 107904 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 236363 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 680001 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 496855 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 616915 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 266341 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 83166 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 182101 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 572 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 120636 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 60630 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 73252 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 53291 | - | - |
| Agilent Whole Rat Genome Microarray 4x44K v3 | A_44_P355861 | - | - |
| Agilent Whole Rat Genome Microarray 4x44K v3 | A_44_P355861 | - | - |
| Agilent SurePrint G3 Rat GE 8x60K Microarray | A_44_P355861 | - | - |
| SurePrint G3 Rat Gene Expression Microarray G4131A | A_44_P457625 | - | - |
| SurePrint G3 Rat Gene Expression Microarray G4131A | A_44_P355861 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 50437 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 44034 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 26885 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 76337 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 1971 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 102644 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 83597 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Rat Gene 1.0 ST Array | 10921963 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10921952 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10921959 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10921966 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10921962 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10921958 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10921954 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10921960 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10921964 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10921957 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10921955 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10921961 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10921965 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10921953 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10921967 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10921956 | - | - |
| Affymetrix Rat Gene 1.1 ST Array | 10921951 | - | - |
| Rat Genome 230 2.0 Array | 1379269_at | - | - |
| Rat Genome 230 2.0 Array | 1393705_PM_at | - | - |
| Rat Genome 230 2.0 Array | 1393705_at | - | - |
| Rat Genome 230 2.0 Array | 1379269_PM_at | - | - |
| Affymetrix Rat Clariom S Assay | PSR0900002714.rn.1 | TC0900000300.rn.1 | RTA TC |
| Affymetrix Rat Clariom S Assay | PSR0900002720.rn.1 | TC0900000300.rn.1 | RTA TC |
| Affymetrix Rat Clariom S Assay | PSR0900002709.rn.1 | TC0900000300.rn.1 | RTA TC |
| Affymetrix Rat Clariom S Assay | PSR0900002716.rn.1 | TC0900000300.rn.1 | RTA TC |
| Affymetrix Rat Clariom S Assay | PSR0900002718.rn.1 | TC0900000300.rn.1 | RTA TC |
| Affymetrix Rat Clariom S Assay | PSR0900002708.rn.1 | TC0900000300.rn.1 | RTA TC |
| Affymetrix Rat Clariom S Assay | PSR0900002715.rn.1 | TC0900000300.rn.1 | RTA TC |
| Affymetrix Rat Clariom S Assay | PSR0900002719.rn.1 | TC0900000300.rn.1 | RTA TC |
| Affymetrix Rat Clariom S Assay | PSR0900002717.rn.1 | TC0900000300.rn.1 | RTA TC |
| Affymetrix Rat Clariom S Assay | PSR0900002711.rn.1 | TC0900000300.rn.1 | RTA TC |
| Affymetrix Rat Clariom S Assay | PSR0900002713.rn.1 | TC0900000300.rn.1 | RTA TC |
| Affymetrix Rat Clariom S Assay | PSR0900002722.rn.1 | TC0900000300.rn.1 | RTA TC |
| Affymetrix Rat Clariom S Assay | PSR0900002723.rn.1 | TC0900000300.rn.1 | RTA TC |
| Affymetrix Rat Clariom S Assay | PSR0900002712.rn.1 | TC0900000300.rn.1 | RTA TC |
| Affymetrix Rat Clariom S Assay | PSR0900002724.rn.1 | TC0900000300.rn.1 | RTA TC |
| Affymetrix Rat Clariom S Assay | PSR0900002721.rn.1 | TC0900000300.rn.1 | RTA TC |
| Affymetrix Rat Clariom S Assay | PSR0900002726.rn.1 | TC0900000300.rn.1 | RTA TC |
| Rat Expression Set 230 | 1379269_PM_at | - | - |
| Rat Expression Set 230 | 1393705_PM_at | - | - |
| Rat Expression Set 230 | 1393705_at | - | - |
| Rat Expression Set 230 | 1379269_at | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10921951 | - | - |
| Rat Genome U34 Set | rc_AI010435_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Rat Genome Survey Microarray | 20994109 | - | - |

Cd2ap
Genomic Map