Assay Details
| Gene Symbol: LOC299271 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_181630.2 | NP_853661.1 | 3 - 4 | 1138 | 87 | |
| GenBank mRNA | AY113703.1 | - | 3 - 4 | 1138 | 87 | |
More Information
Updated Mapping Information
Set Membership: |
|

LOC299271
Genomic Map