Assay Details
| Gene Symbol: Rps27 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_053597.3 | NP_446049.1 | 3 - 4 | 257 | 61 | |
| GenBank mRNA | AF184893.1 | - | 3 - 4 | 251 | 61 | |
| BC061539.1 | - | 261 | 6918361 | 61 | ||
| FQ209846.1 | - | 3 - 4 | 257 | 61 | ||
| FQ210151.1 | - | 3 - 4 | 256 | 61 | ||
| FQ217607.1 | - | 3 - 4 | 259 | 61 | ||
| FQ217624.1 | - | 3 - 4 | 257 | 61 | ||
| FQ218228.1 | - | 3 - 4 | 258 | 61 | ||
| FQ218581.1 | - | 3 - 4 | 257 | 61 | ||
| FQ221131.1 | - | 3 - 4 | 257 | 61 | ||
| FQ221294.1 | - | 3 - 4 | 257 | 61 | ||
| FQ221565.1 | - | 3 - 4 | 257 | 61 | ||
| FQ221645.1 | - | 3 - 4 | 256 | 61 | ||
| FQ221664.1 | - | 3 - 4 | 257 | 61 | ||
| FQ221723.1 | - | 3 - 4 | 256 | 61 | ||
| FQ221727.1 | - | 3 - 4 | 254 | 61 | ||
| FQ221743.1 | - | 3 - 4 | 257 | 61 | ||
| FQ221809.1 | - | 3 - 4 | 257 | 61 | ||
| FQ221911.1 | - | 3 - 4 | 257 | 61 | ||
| FQ221975.1 | - | 3 - 4 | 258 | 61 | ||
| FQ222049.1 | - | 3 - 4 | 257 | 61 | ||
| FQ222054.1 | - | 3 - 4 | 258 | 61 | ||
| FQ222270.1 | - | 3 - 4 | 258 | 61 | ||
| FQ222348.1 | - | 4 - 5 | 374 | 61 | ||
| FQ222848.1 | - | 3 - 4 | 257 | 61 | ||
| FQ222865.1 | - | 3 - 4 | 257 | 61 | ||
| FQ222911.1 | - | 3 - 4 | 257 | 61 | ||
| FQ223072.1 | - | 3 - 4 | 257 | 61 | ||
| FQ223291.1 | - | 3 - 4 | 256 | 61 | ||
| FQ223549.1 | - | 3 - 4 | 257 | 61 | ||
| FQ223862.1 | - | 3 - 4 | 257 | 61 | ||
| FQ224473.1 | - | 3 - 4 | 257 | 61 | ||
| FQ228507.1 | - | 3 - 4 | 258 | 61 | ||
| FQ228599.1 | - | 3 - 4 | 257 | 61 | ||
| FQ228823.1 | - | 3 - 4 | 257 | 61 | ||
| FQ229526.1 | - | 3 - 4 | 257 | 61 | ||
| Gene Symbol: LOC100362987 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002730094.4 | XP_002730140.1 | 1 | 224 | 61 | |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Rps27
Genomic Map