Assay Details
| Gene Symbol: Ap2b1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_080583.1 | NP_542150.1 | 16 - 17 | 2324 | 63 | |
| XM_008767935.1 | XP_008766157.1 | 17 - 18 | 3081 | 63 | ||
| XM_008767936.1 | XP_008766158.1 | 16 - 17 | 2295 | 63 | ||
| XM_008767937.1 | XP_008766159.1 | 15 - 16 | 2210 | 63 | ||
| XM_008767938.1 | XP_008766160.1 | 15 - 16 | 2227 | 63 | ||
| XM_008767939.1 | XP_008766161.1 | 15 - 16 | 3021 | 63 | ||
| GenBank mRNA | BC103481.1 | - | 17 - 18 | 2293 | 7929959 | 63 |
| M34176.1 | - | 15 - 16 | 2214 | 63 | ||
| M77246.1 | - | 16 - 17 | 2324 | 63 | ||
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Ap2b1
Genomic Map