Assay Details
| Gene Symbol: Rpl15 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_139114.3 | NP_620814.1 | 2 - 3 | 244 | 76 | |
| GenBank mRNA | BC078724.1 | - | 1 - 2 | 183 | 7125608 | 76 |
| FQ210520.1 | - | 2 - 3 | 199 | 76 | ||
| FQ217417.1 | - | 2 - 3 | 200 | 76 | ||
| FQ218767.1 | - | 2 - 3 | 199 | 76 | ||
| FQ221301.1 | - | 2 - 3 | 212 | 76 | ||
| FQ221436.1 | - | 2 - 3 | 201 | 76 | ||
| FQ221541.1 | - | 2 - 3 | 200 | 76 | ||
| FQ221627.1 | - | 2 - 3 | 203 | 76 | ||
| FQ221775.1 | - | 2 - 3 | 200 | 76 | ||
| FQ221788.1 | - | 2 - 3 | 199 | 76 | ||
| FQ221816.1 | - | 2 - 3 | 200 | 76 | ||
| FQ222173.1 | - | 2 - 3 | 200 | 76 | ||
| FQ222227.1 | - | 2 - 3 | 203 | 76 | ||
| FQ222322.1 | - | 2 - 3 | 199 | 76 | ||
| FQ222450.1 | - | 2 - 3 | 213 | 76 | ||
| FQ222725.1 | - | 2 - 3 | 200 | 76 | ||
| FQ224390.1 | - | 2 - 3 | 199 | 76 | ||
| FQ228660.1 | - | 1 - 2 | 188 | 76 | ||
| FQ230911.1 | - | 2 - 3 | 203 | 76 | ||
| X78167.1 | - | 1 - 2 | 181 | 76 | ||
| Gene Symbol: RGD1565131 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_006248902.2 | XP_006248964.1 | 1 | 195 | 76 | |
| XM_008769031.1 | XP_008767253.1 | 1 | 195 | 76 | ||
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Rpl15
Genomic Map