Assay Details
| Gene Symbol: Hnrnpa2b1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001104613.1 | NP_001098083.1 | 8 - 9 | 875 | 96 | |
| GenBank mRNA | AB006815.1 | - | 7 - 8 | 839 | 96 | |
| AB006816.1 | - | 8 - 9 | 875 | 96 | ||
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Hnrnpa2b1
Genomic Map