Assay Details
| Gene Symbol: Pknox2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001271278.1 | NP_001258207.1 | 8 - 9 | 1013 | 57 | |
| NM_001271279.1 | NP_001258208.1 | 9 - 10 | 1145 | 57 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Pknox2
Genomic Map