Assay Details
| Gene Symbol: Kctd13 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_198736.1 | NP_942031.1 | 1 - 2 | 424 | 90 | |
| GenBank mRNA | AY305029.1 | - | 1 - 2 | 424 | 90 | |
More Information
Updated Mapping Information
Set Membership: |
|

Kctd13
Genomic Map