Assay Details
| Gene Symbol: Ubl5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001048243.2 | NP_001041708.2 | 1 - 2 | 108 | 83 | |
| GenBank mRNA | CB713235.1 | - | 1 - 2 | 108 | 83 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the NM_001048243.2 transcripts.
Set Membership: |
|

Ubl5
Genomic Map