Assay Details
| Gene Symbol: Chpf2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001106574.1 | NP_001100044.1 | 2 - 3 | 2156 | 56 | |
| XM_006235890.2 | XP_006235952.1 | 3 - 4 | 1313 | 56 | ||
| XM_008762633.1 | XP_008760855.1 | 3 - 4 | 1335 | 56 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Chpf2
Genomic Map