Assay Details
| Gene Symbol: Hic2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001105862.1 | NP_001099332.1 | 2 - 3 | 248 | 92 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the NM_001105862.1 transcripts.
Set Membership: |
|
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Whole Rat Genome Microarray 4x44K v2 | A_44_P366251 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 614705 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 841897 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 674098 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 413772 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 188823 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 435071 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 913969 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 292191 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 581305 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 965452 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 467980 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 928438 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 667852 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 523926 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 348057 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 52186 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 329539 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 867031 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 640230 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 669272 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 29482 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 49190 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 106630 | - | - |
| Agilent Whole Rat Genome Microarray 4x44K v3 | A_42_P738438 | - | - |
| Agilent Whole Rat Genome Microarray 4x44K v3 | A_44_P366251 | - | - |
| Agilent Whole Rat Genome Microarray 4x44K v3 | A_42_P738438 | - | - |
| Agilent Whole Rat Genome Microarray 4x44K v3 | A_44_P366251 | - | - |
| Agilent SurePrint G3 Rat GE 8x60K Microarray | A_42_P738438 | - | - |
| Agilent SurePrint G3 Rat GE 8x60K Microarray | A_44_P366251 | - | - |
| SurePrint G3 Rat Gene Expression Microarray G4131A | A_44_P366251 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 1471 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Rat Gene 1.0 ST Array | 10755786 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10755781 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10755787 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10755783 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10755782 | - | - |
| Affymetrix Rat Gene 1.1 ST Array | 10755780 | - | - |
| Rat Genome 230 2.0 Array | 1390030_at | - | - |
| Rat Genome 230 2.0 Array | 1390030_PM_at | - | - |
| Affymetrix HT Rat Focus Array | 1390030_PM_at | - | - |
| Affymetrix HT Rat Focus Array | 1390030_at | - | - |
| Affymetrix Rat Clariom S Assay | PSR1100013312.rn.1 | TC1100002015.rn.1 | RTA TC |
| Affymetrix Rat Clariom S Assay | PSR1100013313.rn.1 | TC1100002015.rn.1 | RTA TC |
| Affymetrix Rat Clariom S Assay | PSR1100013311.rn.1 | TC1100002015.rn.1 | RTA TC |
| Rat Expression Set 230 | 1390030_at | - | - |
| Rat Expression Set 230 | 1390030_PM_at | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10755780 | - | - |
| Rat Genome U34 Set | rc_AI231432_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Rat Genome Survey Microarray | 21647419 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Illumina ratRef-12 v1.0 expression beadchip | ILMN_1348919 | - | - |

Hic2
Genomic Map