Assay Details
| Gene Symbol: Ppib | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_022536.2 | NP_071981.1 | 3 - 4 | 333 | 91 | |
| GenBank mRNA | AF071225.1 | - | 3 - 4 | 333 | 91 | |
| BC061971.1 | - | 3 - 4 | 362 | 5623342 | 91 | |
| FQ211358.1 | - | 3 - 4 | 337 | 91 | ||
| FQ212031.1 | - | 3 - 4 | 362 | 91 | ||
| FQ219792.1 | - | 3 - 4 | 371 | 91 | ||
| FQ219926.1 | - | 3 - 4 | 383 | 91 | ||
| FQ221020.1 | - | 3 - 4 | 386 | 91 | ||
| FQ221248.1 | - | 3 - 4 | 381 | 91 | ||
| FQ222696.1 | - | 3 - 4 | 495 | 91 | ||
| FQ223162.1 | - | 3 - 4 | 380 | 91 | ||
| FQ229174.1 | - | 3 - 4 | 383 | 91 | ||
| FQ229214.1 | - | 3 - 4 | 377 | 91 | ||
| FQ230004.1 | - | 387 | 91 | |||
| FQ232077.1 | - | 3 - 4 | 372 | 91 | ||
| FQ232920.1 | - | 3 - 4 | 386 | 91 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Ppib
Genomic Map