Assay Details
| Gene Symbol: Phactr2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_214458.1 | NP_999623.1 | 1 - 2 | 63 | 73 | |
| XM_008758610.1 | XP_008756832.1 | 1 - 2 | 983 | 73 | ||
| XM_008758611.1 | XP_008756833.1 | 1 - 2 | 981 | 73 | ||
| GenBank mRNA | AY500158.1 | - | 1 - 2 | 63 | 73 | |
More Information
Updated Mapping Information
Set Membership: |
|

Phactr2
Genomic Map