Assay Details
| Gene Symbol: Ephb2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001127319.1 | NP_001120791.1 | 3 - 4 | 810 | 65 | |
| XM_008764244.1 | XP_008762466.1 | 2 - 3 | 838 | 65 | ||
| XM_008764245.1 | XP_008762467.1 | 3 - 4 | 6532 | 65 | ||
| XM_008764246.1 | XP_008762468.1 | 2 - 3 | 838 | 65 | ||
| XM_008764247.1 | XP_008762469.1 | 2 - 3 | 838 | 65 | ||
| XM_008764248.1 | XP_008762470.1 | 3 - 4 | 792 | 65 | ||
| XM_008764249.1 | XP_008762471.1 | 3 - 4 | 792 | 65 | ||
| XM_008764250.1 | XP_008762472.1 | 3 - 4 | 792 | 65 | ||
| GenBank mRNA | DQ011670.1 | - | 3 - 4 | 810 | 65 | |
More Information
Updated Mapping Information
Set Membership: |
|
Panther Classification:
Gene Ontology Categories:
Function(s)
Process(es)
cell morphogenesis
angiogenesis
urogenital system development
protein phosphorylation
transmembrane receptor protein tyrosine kinase signaling pathway
axon guidance
axonal fasciculation
learning
biological_process
organ morphogenesis
phosphorylation
peptidyl-tyrosine phosphorylation
optic nerve morphogenesis
central nervous system projection neuron axonogenesis
corpus callosum development
retinal ganglion cell axon guidance
inner ear morphogenesis
ephrin receptor signaling pathway
regulation of neuronal synaptic plasticity
positive regulation of long-term neuronal synaptic plasticity
camera-type eye morphogenesis
regulation of axonogenesis
negative regulation of axonogenesis
regulation of body fluid levels
positive regulation of synapse assembly
palate development
dendritic spine development
dendritic spine morphogenesis
commissural neuron axon guidance
molecular_function
protein tyrosine kinase activity
receptor activity
ephrin receptor activity
transmembrane-ephrin receptor activity
receptor binding
ATP binding
axon guidance receptor activity
identical protein binding
cell morphogenesis
angiogenesis
urogenital system development
protein phosphorylation
transmembrane receptor protein tyrosine kinase signaling pathway
axon guidance
axonal fasciculation
learning
biological_process
organ morphogenesis
phosphorylation
peptidyl-tyrosine phosphorylation
optic nerve morphogenesis
central nervous system projection neuron axonogenesis
corpus callosum development
retinal ganglion cell axon guidance
inner ear morphogenesis
ephrin receptor signaling pathway
regulation of neuronal synaptic plasticity
positive regulation of long-term neuronal synaptic plasticity
camera-type eye morphogenesis
regulation of axonogenesis
negative regulation of axonogenesis
regulation of body fluid levels
positive regulation of synapse assembly
palate development
dendritic spine development
dendritic spine morphogenesis
commissural neuron axon guidance
molecular_function
protein tyrosine kinase activity
receptor activity
ephrin receptor activity
transmembrane-ephrin receptor activity
receptor binding
ATP binding
axon guidance receptor activity
identical protein binding
MicroArray Probes:
Agilent
Affymetrix
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Whole Rat Genome Microarray 4x44K v2 | A_64_P030372 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 470869 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 154456 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 692001 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 930440 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 271676 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 251195 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 285753 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 10044 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 483587 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 182100 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 764638 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 331704 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 113853 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 596426 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 195472 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 376226 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 577045 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 484703 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 461947 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 200356 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 453814 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 687184 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 149990 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 652533 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 596560 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 961907 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 552597 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 579974 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 320955 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 640373 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 935930 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 790511 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 580020 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 220783 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 966191 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 265213 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 258356 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 194729 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 240972 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 695031 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 401124 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 184837 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 676786 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 893416 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 208156 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 389636 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 392475 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 172664 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 321626 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 737907 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 876610 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 920533 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 894582 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 647705 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 481434 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 459546 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 389950 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 516526 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 797757 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 80241 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 945555 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 946553 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 395495 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 211845 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 680338 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 869444 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 731175 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 281969 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 872715 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 102049 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 433054 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 946096 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 55184 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 814734 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 719502 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 178998 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 930002 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 916567 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 636611 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 122808 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 801475 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 730860 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 90383 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 137894 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 309198 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 396039 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 631260 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 953846 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 748972 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 441571 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 419409 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 503196 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 90843 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 395140 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 798597 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 955114 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 660415 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 373105 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 666105 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 738226 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 765771 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 33922 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 226031 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 334867 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 201722 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 857172 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 392075 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 259257 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 181231 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 143502 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 583897 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 3050 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 171642 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 690722 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 407582 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 16886 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 272075 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 530677 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 973652 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 460133 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 42918 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 813046 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 19593 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 524963 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 5815 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 718311 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 106871 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 951870 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 565899 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 473462 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 473975 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 403150 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 174971 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 647814 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 619875 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 708985 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 201082 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 637003 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 354709 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 171564 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 55673 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 69990 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 32579 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 91850 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 7418 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 161253 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 22922 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 18616 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 163817 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 114925 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 71681 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 106192 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 109715 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 21250 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 42181 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 158172 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 162372 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 86708 | - | - |
| Agilent Whole Rat Genome Microarray 4x44K v3 | A_64_P030373 | - | - |
| Agilent Whole Rat Genome Microarray 4x44K v3 | A_64_P030373 | - | - |
| Agilent SurePrint G3 Rat GE 8x60K Microarray | A_64_P030373 | - | - |
| SurePrint G3 Rat Gene Expression Microarray G4131A | A_44_P342858 | - | - |
| SurePrint G3 Rat Gene Expression Microarray G4131A | A_44_P538695 | - | - |
| SurePrint G3 Rat Gene Expression Microarray G4131A | A_44_P480780 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 91671 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 46133 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 59327 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 16950 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Rat Gene 1.0 ST Array | 10880723 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10880713 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10880712 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10880714 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10880725 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10880715 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10880716 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10880719 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10880722 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10880721 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10880724 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10880711 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10880720 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10880717 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10880718 | - | - |
| Affymetrix Rat Gene 1.1 ST Array | 10880710 | - | - |
| Rat Genome 230 2.0 Array | 1394660_at | - | - |
| Rat Genome 230 2.0 Array | 1383693_PM_at | - | - |
| Rat Genome 230 2.0 Array | 1394660_PM_at | - | - |
| Rat Genome 230 2.0 Array | 1383693_at | - | - |
| Rat Genome 230 2.0 Array | 1383143_at | - | - |
| Rat Genome 230 2.0 Array | 1383143_PM_at | - | - |
| Affymetrix HT Rat Focus Array | 1383143_at | - | - |
| Affymetrix HT Rat Focus Array | 1383693_at | - | - |
| Affymetrix HT Rat Focus Array | 1383693_PM_at | - | - |
| Affymetrix HT Rat Focus Array | 1383143_PM_at | - | - |
| Affymetrix Rat Clariom S Assay | PSR0500028709.rn.1 | TC0500003821.rn.1 | RTA TC |
| Rat Expression Set 230 | 1383143_at | - | - |
| Rat Expression Set 230 | 1383693_PM_at | - | - |
| Rat Expression Set 230 | 1383693_at | - | - |
| Rat Expression Set 230 | 1383143_PM_at | - | - |
| Rat Expression Set 230 | 1394660_at | - | - |
| Rat Expression Set 230 | 1394660_PM_at | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10880710 | - | - |
| Rat Genome U34 Set | rc_AA955955_at | - | - |
| Rat Genome U34 Set | rc_AI070837_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Illumina ratRef-12 v1.0 expression beadchip | ILMN_1351610 | - | - |

Ephb2
Genomic Map