Assay Details
| Gene Symbol: Rbbp4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001107912.2 | NP_001101382.1 | 3 - 4 | 315 | 76 | |
| GenBank mRNA | BC168994.1 | - | 3 - 4 | 315 | 8362559 | 76 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Rbbp4
Genomic Map