Assay Details
| Gene Symbol: Cdkn2aip | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_006253147.2 | XP_006253209.1 | 2 - 3 | 588 | 108 | |
| GenBank mRNA | BC098694.1 | - | 2 - 3 | 482 | 108 | |
More Information
Set Membership: |
|

Cdkn2aip
Genomic Map