Assay Details
| Gene Symbol: Sdcbp | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_031986.1 | NP_114192.1 | 7 - 8 | 829 | 68 | |
| GenBank mRNA | AF248548.1 | - | 7 - 8 | 829 | 68 | |
| AJ292243.1 | - | 6 - 7 | 772 | 68 | ||
| BC064651.1 | - | 7 - 8 | 797 | 5598152 | 68 | |
More Information
Updated Mapping Information
Set Membership: |
|

Sdcbp
Genomic Map