Assay Details
| Gene Symbol: Snx12 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001108817.2 | NP_001102287.2 | 3 - 4 | 459 | 72 | |
| XM_006257078.1 | XP_006257140.1 | 3 - 4 | 528 | 72 | ||
| XM_006257079.1 | XP_006257141.1 | 3 - 4 | 528 | 72 | ||
| GenBank mRNA | BP500254.1 | - | 3 - 4 | 465 | 72 | |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Snx12
Genomic Map