Assay Details
| Gene Symbol: Ppp1r16b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001191072.1 | NP_001178001.1 | 10 - 11 | 1378 | 58 | |
| XM_008762383.1 | XP_008760605.1 | 10 - 11 | 1462 | 58 | ||
| XM_008762384.1 | XP_008760606.1 | 10 - 11 | 2253 | 58 | ||
| XM_008762385.1 | XP_008760607.1 | 10 - 11 | 2641 | 58 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Ppp1r16b
Genomic Map