Assay Details
| Gene Symbol: Ube2n | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_053928.2 | NP_446380.1 | 1 - 2 | 373 | 101 | |
| GenBank mRNA | BC090072.1 | - | 1 - 2 | 373 | 7113593 | 101 |
More Information
Additional Information:
This assay is in the 5'UTR region of the BC090072.1,NM_053928.2 transcripts.
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Ube2n
Genomic Map