Assay Details
| Gene Symbol: Rnf145 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001105778.2 | NP_001099248.1 | 2 - 3 | 304 | 61 | |
| GenBank mRNA | BC168651.1 | - | 2 - 3 | 304 | 9033412 | 61 |
More Information
Updated Mapping Information
Set Membership: |
|

Rnf145
Genomic Map