Assay Details
| Gene Symbol: LOC100911453 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003749373.3 | XP_003749421.1 | 2 - 3 | 501 | 93 | |
| Gene Symbol: Wdr77 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001008771.3 | NP_001008771.2 | 2 - 3 | 395 | 93 | |
| GenBank mRNA | AY321348.1 | - | 3 - 4 | 338 | 93 | |
| BC097367.1 | - | 2 - 3 | 346 | 7443204 | 93 | |
| CB581439.1 | - | 407 | 93 | |||
| FQ213175.1 | - | 2 - 3 | 490 | 93 | ||
| FQ226328.1 | - | 2 - 3 | 390 | 93 | ||
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect transcript(s) from off target gene(s). Please contact Tech Support if you need more information.
Set Membership: |
|

LOC100911453
Genomic Map