Assay Details
| Gene Symbol: Eif1b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001106867.1 | NP_001100337.1 | 1 - 2 | 206 | 61 | |
| GenBank mRNA | BC169125.1 | - | 1 - 2 | 126 | 6919533 | 61 |
| FQ211778.1 | - | 1 - 2 | 209 | 61 | ||
| FQ214080.1 | - | 1 - 2 | 219 | 61 | ||
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Eif1b
Genomic Map