Assay Details
| Gene Symbol: Rqcd1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001009357.1 | NP_001009357.1 | 6 - 7 | 1052 | 110 | |
| GenBank mRNA | BC087134.1 | - | 6 - 7 | 1052 | 7122272 | 110 |
More Information
Updated Mapping Information
Set Membership: |
|

Rqcd1
Genomic Map