Assay Details
| Gene Symbol: Ankzf1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001014219.1 | NP_001014241.1 | 1 - 2 | 445 | 106 | |
| XM_006245248.2 | XP_006245310.1 | 1 - 2 | 498 | 106 | ||
| GenBank mRNA | BC081973.1 | - | 1 - 2 | 445 | 7126276 | 106 |
More Information
Updated Mapping Information
Set Membership: |
|

Ankzf1
Genomic Map