Assay Details
| Gene Symbol: Spata3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001108805.1 | NP_001102275.1 | 3 - 4 | 762 | 80 | |
| XM_006245411.2 | XP_006245473.1 | 2 - 3 | 569 | 80 | ||
| XM_006245412.2 | XP_006245474.1 | 2 - 3 | 566 | 80 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Spata3
Genomic Map