Assay Details
| Gene Symbol: Ppif | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_172243.1 | NP_758443.1 | 1 - 2 | 222 | 103 | |
| GenBank mRNA | BC086977.1 | - | 1 - 2 | 255 | 7193156 | 103 |
| U68544.1 | - | 1 - 2 | 222 | 103 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Ppif
Genomic Map