Assay Details
| Gene Symbol: Rbmx | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001025663.1 | NP_001020834.1 | 3 - 4 | 340 | 65 | |
| XM_006257680.2 | XP_006257742.1 | 3 - 4 | 421 | 65 | ||
| XM_006257681.2 | XP_006257743.1 | 3 - 4 | 421 | 65 | ||
| XM_006257682.2 | XP_006257744.1 | 3 - 4 | 418 | 65 | ||
| XM_006257683.2 | XP_006257745.1 | 3 - 4 | 553 | 65 | ||
| GenBank mRNA | BC097479.1 | - | 3 - 4 | 340 | 7458559 | 65 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Rbmx
Genomic Map