Assay Details
| Gene Symbol: Npy5r | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | U56078.1 | - | 1 | 46 | 65 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the U56078.1 transcripts.
Set Membership: |
|

Npy5r
Genomic Map