Assay Details
| Gene Symbol: Snapc2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001013121.1 | NP_001013139.1 | 2 - 3 | 323 | 79 | |
| XM_008768956.1 | XP_008767178.1 | 2 - 3 | 426 | 79 | ||
| GenBank mRNA | BC079077.1 | - | 2 - 3 | 323 | 7122319 | 79 |
| FQ214278.1 | - | 1 - 2 | 153 | 79 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Snapc2
Genomic Map