Assay Details
| Gene Symbol: Slc22a13 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001126285.1 | NP_001119757.1 | 3 - 4 | 662 | 77 | |
| XM_008766677.1 | XP_008764899.1 | 3 - 4 | 668 | 77 | ||
| XM_008766678.1 | XP_008764900.1 | 3 - 4 | 668 | 77 | ||
| GenBank mRNA | BC166511.1 | - | 3 - 4 | 662 | 7128568 | 77 |
More Information
Updated Mapping Information
Set Membership: |
|

Slc22a13
Genomic Map