Assay Details
| Gene Symbol: Ephb3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001105868.1 | NP_001099338.1 | 2 - 3 | 572 | 57 | |
| XM_006248561.2 | XP_006248623.1 | 2 - 3 | 700 | 57 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Ephb3
Genomic Map