Assay Details
| Gene Symbol: Ypel1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002727914.4 | XP_002727960.1 | 1 - 2 | 185 | 122 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_002727914.4 transcripts.
Set Membership: |
|

Ypel1
Genomic Map