Assay Details
| Gene Symbol: LOC100912455 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003751081.3 | XP_003751129.1 | 1 - 2 | 735 | 129 | |
| XM_006248554.2 | XP_006248616.1 | 1 - 2 | 735 | 129 | ||
| Gene Symbol: Clcn2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_017137.1 | NP_058833.1 | 242 | 129 | ||
| XM_006248562.2 | XP_006248624.1 | 735 | 129 | |||
| XM_006248563.2 | XP_006248625.1 | 735 | 129 | |||
| GenBank mRNA | X64139.1 | - | 242 | 129 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the NM_017137.1, X64139.1, XM_003751081.3, XM_006248554.2, XM_006248562.2, XM_006248563.2 transcripts.
Set Membership: |
|

LOC100912455
Genomic Map