Assay Details
| Gene Symbol: Pkp2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001100499.1 | NP_001093969.1 | 2361 | 68 | ||
| GenBank mRNA | BC092602.1 | - | 7 - 8 | 968 | 68 | |
More Information
Set Membership: |
|
Panther Classification:
Gene Ontology Categories:
Function(s)
Process(es)
desmosome assembly
heart development
negative regulation of cell proliferation
gap junction assembly
single organismal cell-cell adhesion
negative regulation of cell migration
adherens junction maintenance
intermediate filament bundle assembly
maintenance of organ identity
ventricular cardiac muscle tissue morphogenesis
lipid homeostasis
cardiac muscle cell action potential
cardiac muscle cell action potential involved in contraction
ventricular cardiac muscle cell action potential
cell-cell signaling involved in cardiac conduction
bundle of His cell to Purkinje myocyte communication
regulation of heart rate by cardiac conduction
regulation of bicellular tight junction assembly
protein kinase C binding
protein binding
intermediate filament binding
protein complex scaffold
desmosome assembly
heart development
negative regulation of cell proliferation
gap junction assembly
single organismal cell-cell adhesion
negative regulation of cell migration
adherens junction maintenance
intermediate filament bundle assembly
maintenance of organ identity
ventricular cardiac muscle tissue morphogenesis
lipid homeostasis
cardiac muscle cell action potential
cardiac muscle cell action potential involved in contraction
ventricular cardiac muscle cell action potential
cell-cell signaling involved in cardiac conduction
bundle of His cell to Purkinje myocyte communication
regulation of heart rate by cardiac conduction
regulation of bicellular tight junction assembly
protein kinase C binding
protein binding
intermediate filament binding
protein complex scaffold
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Whole Rat Genome Microarray 4x44K v2 | A_44_P999507 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 400865 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 543562 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 840066 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 664808 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 382105 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 270691 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 346543 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 349701 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 552965 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 202239 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 155979 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 685173 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 370165 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 368082 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 429755 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 53738 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 208896 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 646668 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 256972 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 437720 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 22320 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 7520 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 7332 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 71209 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 123800 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 30042 | - | - |
| Agilent Whole Rat Genome Microarray 4x44K v3 | A_44_P999507 | - | - |
| Agilent Whole Rat Genome Microarray 4x44K v3 | A_44_P999507 | - | - |
| Agilent SurePrint G3 Rat GE 8x60K Microarray | A_44_P999507 | - | - |
| SurePrint G3 Rat Gene Expression Microarray G4131A | A_44_P319349 | - | - |
| SurePrint G3 Rat Gene Expression Microarray G4131A | A_44_P746697 | - | - |
| SurePrint G3 Rat Gene Expression Microarray G4131A | A_44_P999507 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 68071 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Rat Gene 1.0 ST Array | 10752584 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10752583 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10752579 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10752585 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10752582 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10752581 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10752578 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10752580 | - | - |
| Affymetrix Rat Gene 1.1 ST Array | 10752576 | - | - |
| Rat Genome 230 2.0 Array | 1388539_PM_at | - | - |
| Rat Genome 230 2.0 Array | 1377475_PM_at | - | - |
| Rat Genome 230 2.0 Array | 1388539_at | - | - |
| Rat Genome 230 2.0 Array | 1377475_at | - | - |
| Affymetrix HT Rat Focus Array | 1377475_PM_at | - | - |
| Affymetrix HT Rat Focus Array | 1388539_PM_at | - | - |
| Affymetrix HT Rat Focus Array | 1377475_at | - | - |
| Affymetrix HT Rat Focus Array | 1388539_at | - | - |
| Rat Expression Set 230 | 1388539_at | - | - |
| Rat Expression Set 230 | 1377475_PM_at | - | - |
| Rat Expression Set 230 | 1388539_PM_at | - | - |
| Rat Expression Set 230 | 1377475_at | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10752576 | - | - |
| Rat Genome U34 Set | rc_AA925104_at | - | - |
| Rat Genome U34 Set | rc_AI071117_at | - | - |
| Rat Genome U34 Set | rc_AI072896_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Rat Genome Survey Microarray | 22109943 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Illumina ratRef-12 v1.0 expression beadchip | ILMN_1369107 | - | - |

Pkp2
Genomic Map