Assay Details
| Gene Symbol: Prodh | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001135778.1 | NP_001129250.1 | 12 - 13 | 1545 | 59 | |
| XM_006248642.2 | XP_006248704.2 | 12 - 13 | 1560 | 59 | ||
| XM_006248643.1 | XP_006248705.1 | 13 - 14 | 1487 | 59 | ||
| XM_008768903.1 | XP_008767125.1 | 12 - 13 | 1309 | 59 | ||
| GenBank mRNA | BC168225.1 | - | 7 - 8 | 713 | 59 | |
More Information
Updated Mapping Information
Set Membership: |
|

Prodh
Genomic Map