Assay Details
| Gene Symbol: Tmem191c | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_001059247.5 | XP_001059247.3 | 2 - 3 | 861 | 72 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

Tmem191c| Gene Symbol: Tmem191c | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_001059247.5 | XP_001059247.3 | 2 - 3 | 861 | 72 | |
Set Membership: |
|