Assay Details
| Gene Symbol: Tmem191c | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_001059247.5 | XP_001059247.3 | 8 - 9 | 1294 | 157 | |
| XM_006248711.2 | XP_006248773.1 | 7 - 8 | 1447 | 157 | ||
More Information
Set Membership: |
|

Tmem191c
Genomic Map