Assay Details
| Gene Symbol: Dsg1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_008771993.1 | XP_008770215.1 | 3 | 336 | 84 | |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
This assay is in the 5'UTR region of the XM_008771993.1 transcripts.
This assay is in the 5'UTR region of the XM_008771993.1 transcripts.
Set Membership: |
|

Dsg1
Genomic Map