Assay Details
| Gene Symbol: Pcdhga5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001037137.1 | NP_001032214.1 | 2594 | 95 | ||
| GenBank mRNA | AY574016.1 | - | 2594 | 95 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Pcdhga5
Genomic Map