Assay Details
| Gene Symbol: Gnpda1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001134995.1 | NP_001128467.1 | 4 - 5 | 817 | 76 | |
| GenBank mRNA | BC169009.1 | - | 4 - 5 | 779 | 7312423 | 76 |
More Information
Updated Mapping Information
Set Membership: |
|

Gnpda1
Genomic Map