Assay Details
| Gene Symbol: Brd1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001108103.1 | NP_001101573.1 | 2 - 3 | 1415 | 63 | |
| XM_006242194.2 | XP_006242256.1 | 2 - 3 | 1411 | 63 | ||
| XM_006242196.2 | XP_006242258.1 | 2 - 3 | 1411 | 63 | ||
| XM_006242197.2 | XP_006242259.1 | 2 - 3 | 1411 | 63 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Brd1
Genomic Map