Assay Details
| Gene Symbol: Gpr39 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001100943.1 | NP_001094413.1 | 1 | 828 | 149 | |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|
Panther Classification:
Gene Ontology Categories:
Function(s)
Process(es)
diet induced thermogenesis
G-protein coupled receptor signaling pathway
adult feeding behavior
regulation of lipid metabolic process
positive regulation of insulin secretion
gastric emptying
positive regulation of insulin secretion involved in cellular response to glucose stimulus
regulation of membrane potential
glucose homeostasis
cholesterol homeostasis
positive regulation of release of sequestered calcium ion into cytosol
negative regulation of gastric acid secretion
cellular response to zinc ion
G-protein coupled receptor activity
diet induced thermogenesis
G-protein coupled receptor signaling pathway
adult feeding behavior
regulation of lipid metabolic process
positive regulation of insulin secretion
gastric emptying
positive regulation of insulin secretion involved in cellular response to glucose stimulus
regulation of membrane potential
glucose homeostasis
cholesterol homeostasis
positive regulation of release of sequestered calcium ion into cytosol
negative regulation of gastric acid secretion
cellular response to zinc ion
G-protein coupled receptor activity
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Whole Rat Genome Microarray 4x44K v2 | A_64_P133944 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 189388 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 798375 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 560844 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 87723 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 446255 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 227194 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 153221 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 360341 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 200066 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 789123 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 633007 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 854513 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 897549 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 673401 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 155003 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 718892 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 820268 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 62606 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 615448 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 284255 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 633944 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 665918 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 386040 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 451172 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 49696 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 509112 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 530396 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 683739 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 228445 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 632457 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 406746 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 573071 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 83289 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 628020 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 769499 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 445438 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 117144 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 666811 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 11390 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 391505 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 409447 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 664646 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 321955 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 580949 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 49182 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 786441 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 313306 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 662095 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 682029 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 742887 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 906746 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 116522 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 825762 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 550876 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 791993 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 750213 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 284727 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 719480 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 784074 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 775173 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 696141 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 368469 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 81377 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 75550 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 817546 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 737769 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 112774 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 472120 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 449780 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 450323 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 290565 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 258947 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 697896 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 79016 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 598623 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 571833 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 626513 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 885142 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 604511 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 244473 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 108093 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 775295 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 805265 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 337546 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 75498 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 84965 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 240489 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 230841 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 106497 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 52684 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 645266 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 166040 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 793025 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 19977 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 30884 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 729383 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 212345 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 153441 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 765157 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 504890 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 44287 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 75638 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 873457 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 894501 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 106614 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 281796 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 753316 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 833365 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 395252 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 331233 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 878462 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 534276 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 906365 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 560545 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 957891 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 559264 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 493355 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 356917 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 482664 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 555629 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 431315 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 811382 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 778293 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 46130 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 944114 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 27120 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 261872 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 65134 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 33566 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 895218 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 667378 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 457713 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 485903 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 856684 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 14372 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 724655 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 62827 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 454600 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 229559 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 400947 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 315286 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 729507 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 831604 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 248550 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 709705 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 314665 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 846201 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 863758 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 387335 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 935602 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 298963 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 874057 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 4342 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 4700 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 88674 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 65052 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 107844 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 26765 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 76052 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 179715 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 132076 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 40777 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 175485 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 37953 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 127276 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 144495 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 7167 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 145880 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 127388 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 149721 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 37420 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 32736 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 52258 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 34584 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 59730 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 16283 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 88034 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 162209 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 26153 | - | - |
| Agilent Whole Rat Genome Microarray 4x44K v3 | A_64_P133944 | - | - |
| Agilent Whole Rat Genome Microarray 4x44K v3 | A_64_P133944 | - | - |
| Agilent SurePrint G3 Rat GE 8x60K Microarray | A_64_P133944 | - | - |
| SurePrint G3 Rat Gene Expression Microarray G4131A | A_44_P426512 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 60980 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 34634 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 59760 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 6753 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 49926 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 30425 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 92032 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Rat Gene 1.0 ST Array | 10763605 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10763606 | - | - |
| Affymetrix Rat Gene 1.1 ST Array | 10763604 | - | - |
| Affymetrix Rat Gene 1.1 ST Array | 10767243 | - | - |
| Affymetrix Rat Clariom S Assay | PSR1300000907.rn.1 | TC1300000180.rn.1 | RTA TC |
| Affymetrix Rat Gene 1.0 ST Array | 10763604 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Rat Genome Survey Microarray | 22083218 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Illumina ratRef-12 v1.0 expression beadchip | ILMN_1356331 | - | - |

Gpr39
Genomic Map